EMAIL THIS PAGE TO A FRIEND
International Journal of Oncology
Inhibition of caspase-9 by oridonin, a diterpenoid isolated from Rabdosia rubescens, augments apoptosis in human laryngeal cancer cells.
PMID 26648189