EMAIL THIS PAGE TO A FRIEND
BMC Public Health
Relationship between risk factors for infertility in women and lead, cadmium, and arsenic blood levels: a cross-sectional study from Taiwan.
PMID 26653029