EMAIL THIS PAGE TO A FRIEND
Scientific reports
PTEN stabilizes TOP2A and regulates the DNA decatenation.
PMID 26657567
Abstract
PTEN is a powerful tumor suppressor that antagonizes the cytoplasmic PI3K-AKT pathway and suppresses cellular proliferation. PTEN also plays a role in the maintenance of genomic stability in the nucleus. Here we report that PTEN facilitates DNA decatenation and controls a decatenation checkpoint. Catenations of DNA formed during replication are decatenated by DNA topoisomerase II (TOP2), and this process is actively monitored by a decatenation checkpoint in G2 phase. We found that PTEN deficient cells form ultra-fine bridges (UFBs) during anaphase and these bridges are generated as a result of insufficient decatenation. We show that PTEN is physically associated with a decatenation enzyme TOP2A and that PTEN influences its stability through OTUD3 deubiquitinase. In the presence of PTEN, ubiquitination of TOP2A is inhibited by OTUD3. Deletion or deficiency of PTEN leads to down regulation of TOP2A, dysfunction of the decatenation checkpoint and incomplete DNA decatenation in G2 and M phases. We propose that PTEN controls DNA decatenation to maintain genomic stability and integrity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D7385
Demecolcine, ≥98% (HPLC)
C21H25NO5
CLLS1004 DLD1 Cells PTEN -/-
RDD002
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
C8H18N2O4S
H3375
HEPES, ≥99.5% (titration)
C8H18N2O4S
H4034
HEPES, BioPerformance Certified, ≥99.5% (titration), cell culture tested
C8H18N2O4S
PHG0001
HEPES, PharmaGrade, Manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production, suitable for cell culture
C8H18N2O4S
RES6003H-B7
HEPES, PharmaGrade, Manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production, suitable for cell culture
C8H18N2O4S
H7523
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
C8H18N2O4S
H6147
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
C8H18N2O4S
54457
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
C8H18N2O4S
83264
HEPES buffer solution, 1 M in H2O
C8H18N2O4S
449172
Magnesium chloride, anhydrous, beads, −10 mesh, 99.99% trace metals basis
Cl2Mg
449164
Magnesium chloride, anhydrous, beads, −10 mesh, 99.9% trace metals basis
Cl2Mg
M8266
Magnesium chloride, anhydrous, ≥98%
Cl2Mg
M4880
Magnesium chloride, BioReagent, suitable for insect cell culture, ≥97.0%
Cl2Mg
208337
Magnesium chloride, powder, <200 μm
Cl2Mg
M1028
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Cl2Mg
63069
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Cl2Mg
M8787
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Cl2Mg
63036
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Cl2Mg
68475
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Cl2Mg
00457
Magnesium chloride solution, 0.1 M
Cl2Mg
74255
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
C12H25NaO4S
L3771
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
C12H25NaO4S
L6026
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
C12H25NaO4S
436143
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
C12H25NaO4S
L4509
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
C12H25NaO4S
L5750
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
C12H25NaO4S
71725
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
C12H25NaO4S
71729
Sodium dodecyl sulfate, ≥98.0% (GC)
C12H25NaO4S