EMAIL THIS PAGE TO A FRIEND
The EMBO journal
A hit-and-run heat shock factor governs sustained histone methylation and transcriptional stress memory.
PMID 26657708
Abstract
In nature, plants often encounter chronic or recurring stressful conditions. Recent results indicate that plants can remember a past exposure to stress to be better prepared for a future stress incident. However, the molecular basis of this is poorly understood. Here, we report the involvement of chromatin modifications in the maintenance of acquired thermotolerance (heat stress [HS] memory). HS memory is associated with the accumulation of histone H3 lysine 4 di- and trimethylation at memory-related loci. This accumulation outlasts their transcriptional activity and marks them as recently transcriptionally active. High accumulation of H3K4 methylation is associated with hyper-induction of gene expression upon a recurring HS. This transcriptional memory and the sustained accumulation of H3K4 methylation depend on HSFA2, a transcription factor that is required for HS memory, but not initial heat responses. Interestingly, HSFA2 associates with memory-related loci transiently during the early stages following HS. In summary, we show that transcriptional memory after HS is associated with sustained H3K4 hyper-methylation and depends on a hit-and-run transcription factor, thus providing a molecular framework for HS memory.