EMAIL THIS PAGE TO A FRIEND
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism
The potassium channel KCa3.1 constitutes a pharmacological target for neuroinflammation associated with ischemia/reperfusion stroke.
PMID 26661208
Abstract
Activated microglia/macrophages significantly contribute to the secondary inflammatory damage in ischemic stroke. Cultured neonatal microglia express the K
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV35098
Anti-KCNN4 antibody produced in rabbit, affinity isolated antibody