EMAIL THIS PAGE TO A FRIEND
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism
Environmental enrichment enhances synaptic plasticity by internalization of striatal dopamine transporters.
PMID 26661218
Abstract
Environmental enrichment (EE) with a complex combination of physical, cognitive and social stimulations enhances synaptic plasticity and behavioral function. However, the mechanism remains to be elucidated in detail. We aimed to investigate dopamine-related synaptic plasticity underlying functional improvement after EE. For this, six-week-old CD-1 mice were randomly allocated to EE or standard conditions for two months. EE significantly enhanced behavioral functions such as rotarod and ladder walking tests. In a [