EMAIL THIS PAGE TO A FRIEND
The Journal of antibiotics
Distribution of virginiae butanolides in antibiotic-producing actinomycetes, and identification of the inducing factor from Streptomyces antibioticus as virginiae butanolide A.
PMID 2666375
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SMB00045
Virginiaebutanolide C, ≥90% (LC/MS-ELSD)
C11H20O4