EMAIL THIS PAGE TO A FRIEND
Beilstein journal of organic chemistry
Two strategies for the synthesis of the biologically important ATP analogue ApppI, at a multi-milligram scale.
PMID 26664641
Abstract
Two strategies for the synthesis of the ATP (adenosine triphosphate) analogue ApppI [1-adenosin-5'-yl 3-(3-methylbut-3-enyl)triphosphoric acid diester] (1) are described. ApppI is an active metabolite of the mevalonate pathway and thus is of major biological significance. Chemically synthezised ApppI was purified by using triethylammonium bicarbonate as the counter ion in ion-pair chromatography and characterized by (1)H, (13)C, (31)P NMR and MS spectroscopical methods.