EMAIL THIS PAGE TO A FRIEND
Oncotarget
Metformin increases antitumor activity of MEK inhibitors through GLI1 downregulation in LKB1 positive human NSCLC cancer cells.
PMID 26673006
Abstract
Metformin, widely used as antidiabetic drug, showed antitumoral effects expecially in combination with chemotherapy. Our group recently has demonstrated that metformin and gefitinib are synergistic in LKB1-wild-type NSCLC cells. In these models, metformin as single agent induced an activation and phosphorylation of mitogen-activated-protein-kinase (MAPK) through an increased C-RAF/B-RAF heterodimerization. Since single agent metformin enhances proliferating signals through the RAS/RAF/MAPK pathway, and several MEK inhibitors (MEK-I) demonstrated clinical efficacy in combination with other agents in NSCLC, we tested the effects of metformin plus MEK-I (selumetinib or pimasertib) on proliferation, invasiveness, migration abilities in vitro and in vivo in LKB1 positive NSCLC models harboring KRAS wild type and mutated gene. The combination of metformin with MEK-I showed a strong anti-proliferative and proapoptotic effect in Calu-3, H1299, H358 and H1975 human NSCLC cell lines, independently from the KRAS mutational status. The combination reduced the metastatic behaviour of NSCLC cells, via a downregulation of GLI1 trascritional activity, thus affecting the transition from an epithelial to a mesenchymal phenotype. Metformin and MEK-Is combinations also decreased the production and activity of MMP-2 and MMP-9 by reducing the NF-jB (p65) binding to MMP-2 and MMP-9 promoters. Metformin potentiates the antitumor activity of MEK-Is in human LKB1-wild-type NSCLC cell lines, independently from the KRAS mutational status, through GLI1 downregulation and by reducing the NF-jB (p65)-mediated transcription of MMP-2 and MMP-9.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D9760
L-(−)-Dithiothreitol, ≥95%
C4H10O2S2