EMAIL THIS PAGE TO A FRIEND
Drug design, development and therapy
Tanshinone IIA ameliorates dextran sulfate sodium-induced inflammatory bowel disease via the pregnane X receptor.
PMID 26674743
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EHU081471 MISSION® esiRNA, esiRNA human NR1I2 (esiRNA1)