EMAIL THIS PAGE TO A FRIEND
Chemphyschem : a European journal of chemical physics and physical chemistry
Air-Stable, Near- to Mid-Infrared Emitting Solids of PbTe/CdTe Core-Shell Colloidal quantum dots.
PMID 26676076
Abstract
Light emitters and detectors operating in the near- and mid-infrared spectral regions are important to many applications, such as telecommunications, high-resolution gas analysis, atmospheric pollution monitoring, medical diagnostics, and night vision. Various lead chalcogenides (binary, ternary, and quaternary alloys) in the form of quantum dots (QDs) or quantum wells provide narrow bandgap energies that cover the broad infrared region corresponding to wavelengths of 1-30 μm. Here, we report an inexpensive, all-solution-based synthesis strategy to thin-film solids consisting of 5-16 nm PbTe QDs encapsulated by CdTe shells. Colloidally synthesized PbTe QDs were first converted into core-shell PbTe/CdTe QDs, and then deposited as thin films. The subsequent fusion of the CdTe shells is achieved by ligand removal and annealing in the presence of CdCl2 . Contrary to highly unstable bare PbTe QDs, PbTe/CdTe QD solids exhibit bright and stable near- to mid-infrared emission at wavelengths of 1-3 μm, which is also retained upon prolonged storage at ambient conditions for one year.
Related Materials