EMAIL THIS PAGE TO A FRIEND
Environmental toxicology
Arsenite-induced endoplasmic reticulum-dependent apoptosis through disturbance of calcium homeostasis in HBE cell line.
PMID 26677073
Abstract
Calcium (Ca