EMAIL THIS PAGE TO A FRIEND
OncoTargets and Therapy
Echinacoside induces apoptotic cancer cell death by inhibiting the nucleotide pool sanitizing enzyme MTH1.
PMID 26677335
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
PZ0240
(S)-Crizotinib, ≥98% (HPLC)
C21H22Cl2FN5O
07668
Echinacoside, 98% (HPLC)
C35H46O20