EMAIL THIS PAGE TO A FRIEND
Journal of Analytical Methods in Chemistry
Development of Sensitive and Specific Analysis of Vildagliptin in Pharmaceutical Formulation by Gas Chromatography-Mass Spectrometry.
PMID 26682085