EMAIL THIS PAGE TO A FRIEND
Journal of Nanoscience and Nanotechnology
Breast Cancer Tissue Marked Selectively by Magnetic Nanoparticles in an Experimental Animal Model.
PMID 26682383
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N1517
N-Nitroso-N-methylurea, ISOPAC®
C2H5N3O2
N4766
N-Nitroso-N-methylurea, Bulk package
C2H5N3O2