EMAIL THIS PAGE TO A FRIEND
Journal of Nanoscience and Nanotechnology
A Novel Intelligent Vascular Stent: Preparation and Characterization of Hydrophobic Steel Mesh Stent with Liquid Surface Tension Response.
PMID 26682391