EMAIL THIS PAGE TO A FRIEND
BMC complementary and alternative medicine
Effect of an ethanol extract of Descurainia sophia seeds on Phase I and II drug metabolizing enzymes and P-glycoprotein activity in vitro.
PMID 26683337
Abstract
Descurainia sophia seeds have a variety of pharmacological functions and been widely used in traditional folk medicine. However, their effects on human drug metabolizing enzyme (DME) activities have not been elucidated. The present study investigated the inhibitory effects of an ethanol extract of D. sophia seeds (EEDS) on human Phase I/II (DMEs) and P-glycoprotein (p-gp) in vitro. The enzyme activities of human Phase I (cytochrome P450s, CYPs), Phase II (uridine diphosphate glucuronosyltransferases, UGTs) DMEs, and the drug transporter P-gp were determined in the presence of various concentrations of EEDS using commercially available luminogenic assay systems. The mode of enzyme inhibition and the inhibitory constant (Ki) value of EEDS were graphically determined with Lineweaver-Burk double reciprocal plots and secondary plots, respectively. The enzyme activity assays showed that EEDS moderately inhibited the CYP1A2, CYP2C9, and CYP2C19 isoforms with half maximal inhibitory concentrations (IC50) of 47.3, 25.8, and 38.7xa0μg/mL, respectively. Graphical analyses with Lineweaver-Burk double reciprocal plots and secondary plots indicated that EEDS competitively inhibited CYP2C9 with a Ki value of 19.8xa0μg/mL; however, it inhibited CYP2C9 and CYP2C19 in a mixed mode with Ki values of 5.2, and 11.9xa0μg/mL, respectively. Other Phase I (CYP2C8, CYP2D6, and CYP3A4) and Phase II (UGT1A1 and UGT2B7) enzymes as well as P-gp were weakly or negligibly affected by EEDS with concentrations up to 500xa0μg/mL. EEDS is a selective inhibitor of CYP1A2, CYP2C9, and CYP2C19 with moderate enzymatic inhibition. Clinically, full consideration should be given to a potential toxic adverse effect from a herb-drug interaction when drugs that are particularly susceptible to CYP1A2, CYP2C9, or CYP2C19-mediated metabolism are taken together with EEDS. Characterization of metabolic profiles of specific herbal drugs could help consumers and medical specialists to use them safely as a complementary and alternative medicine.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SRP6071
Cyclophilin G human, recombinant, expressed in E. coli, 95 (SDS-PAGE)