EMAIL THIS PAGE TO A FRIEND
Journal of Hazardous Materials
A REVIEW OF THE CURRENT STATE OF THE ART OF PHYSIOLOGICALLY-BASED TESTS FOR MEASURING HUMAN DERMAL IN VITRO BIOAVAILABILITY OF POLYCYCLIC AROMATIC HYDROCARBONS (PAH) IN SOIL
PMID 26686483