EMAIL THIS PAGE TO A FRIEND
Journal of Ophthalmology
Safety and Biocompatibility of a New High-Density Polyethylene-Based Spherical Integrated Porous Orbital Implant: An Experimental Study in Rabbits.
PMID 26689343
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D35201
N,N′-Dibenzylethylenediamine, 97%
C16H20N2