EMAIL THIS PAGE TO A FRIEND
Nature neuroscience
The indirect pathway of the nucleus accumbens shell amplifies neuropathic pain.
PMID 26691834
Abstract
We examined adaptations in nucleus accumbens (NAc) neurons in mouse and rat peripheral nerve injury models of neuropathic pain. Injury selectively increased excitability of NAc shell indirect pathway spiny projection neurons (iSPNs) and altered their synaptic connectivity. Moreover, injury-induced tactile allodynia was reversed by inhibiting and exacerbated by exciting iSPNs, indicating that they not only participated in the central representation of pain, but gated activity in ascending nociceptive pathways.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B4261
Biocytin, ≥98% (TLC)
C16H28N4O4S