EMAIL THIS PAGE TO A FRIEND
Talanta
Preparation and characterization of magnetic allylamine modified graphene oxide-poly(vinyl acetate-co-divinylbenzene) nanocomposite for vortex assisted magnetic solid phase extraction of some metal ions.
PMID 26695244