EMAIL THIS PAGE TO A FRIEND
Talanta
A new magnetic ion-imprinted polymer as a highly selective sorbent for determination of cobalt in biological and environmental samples.
PMID 26695259
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A8380
5α-Androstan-17β-ol-3-one, ≥97.5%
C19H30O2
45726
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
C2H4O2
W200603
Acetic acid, natural, ≥99.5%, FG
C2H4O2
W200611
Acetic acid, ≥99.5%, FCC, FG
C2H4O2
ARK2183
Acetic acid, glacial, PharmaGrade, USP, JP, Ph Eur, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production.
C2H4O2
338818
Ammonium hydroxide solution, 28% NH3 in H2O, ≥99.99% trace metals basis
H5NO
221228
Ammonium hydroxide solution, ACS reagent, 28.0-30.0% NH3 basis
H5NO
17093
Ammonium hydroxide solution, puriss. p.a. plus, ≥25% NH3 in H2O
H5NO
30501
Ammonium hydroxide solution, puriss. p.a., reag. ISO, reag. Ph. Eur., ~25% NH3 basis
H5NO
05002
Ammonium hydroxide solution, puriss., 30-33% NH3 in H2O
H5NO
05003
Ammonium hydroxide solution, puriss., meets analytical specification of Ph. Eur., 25-30% NH3 basis
H5NO
40205
Ammonium hydroxide solution, ≥25% NH3 in H2O, semiconductor grade PURANAL (Honeywell 17814)
H5NO
01-4362
Ammonium hydroxide solution, 28% NH3 in H2O, suitable for determination of toxic metals
H5NO
2858
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
C2H6O
48075
Ethanol solution, certified reference material, 2000 μg/mL in methanol
C2H6O
E2385
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
C2H6O
792780
Ethyl alcohol, Pure, 200 proof, ACS reagent, meets USP testing specifications, Excise Tax-free, Permit for use required
C2H6O
792799
Ethyl alcohol, Pure, 190 proof, ACS reagent, meets USP testing specifications, Excise Tax-free, Permit for use required
C2H6O
459836
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
C2H6O
493538
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
C2H6O
493511
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
C2H6O
438073
Nitric acid, ACS reagent, 70%
HNO3
225711
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis
HNO3
309079
Nitric acid, red, fuming, HNO3 >90 %
HNO3
84392
Nitric acid, purum p.a., fuming, packed in coated, shock- and leak-protected glass bottle, ≥99% (T)
HNO3
84378
Nitric acid, puriss. p.a., 65% (Hg ≤0.0000005%), ≥65% (T)
HNO3
84380
Nitric acid, puriss. p.a., ≥65% (T)
HNO3
30709
Nitric acid, puriss. p.a., reag. ISO, reag. Ph. Eur., for determinations with dithizone, ≥65%
HNO3
695025
Nitric acid, reagent grade, fuming, >90%
HNO3
695041
Nitric acid, ACS reagent, ≥90.0%
HNO3