EMAIL THIS PAGE TO A FRIEND
Talanta
Reproducible E. coli detection based on label-free SERS and mapping.
PMID 26695290
Abstract
The biosensing for rapid detection of bacteria based on surface-enhanced Raman scattering (SERS) has been widely explored for recent years. It is still a challenge to achieve a high sensitive, reproducible label free detection method for bacteria. In this work, a label-free SERS detection method of Escherichia coli based on incubation with silver colloid was reported. Optimized incubation conditions including shaking speed, time and temperature were used to help construct a rapid SERS method for E. coli analysis. It was found that the enhancement of the Raman signal of E. coli could be achieved to 1.8×10(4) cps (counts per second) with high reproducibility. Three strains of E. coli DSM 1116/498/5695 could be successfully discriminated using such SERS method combining discriminant analysis. Finally, the lowest concentration of E. coli at 1×10(5) cell/mL can be detected by SERS mapping. Thus, our detection method offers higher sensitivity and reproducibility compared to previously reported label free simple-mixing methods, opening an avenue for developing various SERS-based biosensor.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
282979
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
C7H12O2
H1758
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
HCl
84429
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
HCl
H9892
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
HCl
2104
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
HCl
W530574
Hydrochloric acid solution, 32 wt. % in H2O, FCC
HCl
295426
Hydrogen chloride, ReagentPlus®, ≥99%
HCl
705551
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
HCl
379921
Hydroxylamine hydrochloride, 99.995% trace metals basis
H3NO · HCl
431362
Hydroxylamine hydrochloride, 99.999% trace metals basis
H3NO · HCl
255580
Hydroxylamine hydrochloride, ACS reagent, 98.0%
H3NO · HCl
159417
Hydroxylamine hydrochloride, ReagentPlus®, 99%
H3NO · HCl
55460
Hydroxylamine hydrochloride, puriss. p.a., ACS reagent, ≥99.0% (RT)
H3NO · HCl
467804
Hydroxylamine solution, 50 wt. % in H2O, 99.999%
H3NO
438227
Hydroxylamine solution, 50 wt. % in H2O
H3NO
40104-U Methanolic HCl, 3 M HCl in methanol (3N), for GC derivatization, pkg of 1 × 2 L
HCl
438073
Nitric acid, ACS reagent, 70%
HNO3
225711
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis
HNO3
309079
Nitric acid, red, fuming, HNO3 >90 %
HNO3
84392
Nitric acid, purum p.a., fuming, packed in coated, shock- and leak-protected glass bottle, ≥99% (T)
HNO3
84378
Nitric acid, puriss. p.a., 65% (Hg ≤0.0000005%), ≥65% (T)
HNO3
84380
Nitric acid, puriss. p.a., ≥65% (T)
HNO3
30709
Nitric acid, puriss. p.a., reag. ISO, reag. Ph. Eur., for determinations with dithizone, ≥65%
HNO3
695025
Nitric acid, reagent grade, fuming, >90%
HNO3
695041
Nitric acid, ACS reagent, ≥90.0%
HNO3
695033
Nitric acid, ACS reagent, 70%
HNO3
71687
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
HNaO
757527
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
HNaO
72068
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
HNaO
S2770
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
HNaO