EMAIL THIS PAGE TO A FRIEND
Talanta
Multi-residue method for determination of 58 pesticides, pharmaceuticals and personal care products in water using solvent demulsification dispersive liquid-liquid microextraction combined with liquid chromatography-tandem mass spectrometry.
PMID 26695317
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
297887
1-Octanol, anhydrous, ≥99%
C8H18O
W280003
1-Octanol, ≥98%, FCC, FG
C8H18O
W280011
1-Octanol, natural, ≥98%, FCC
C8H18O
49083
2,4-D, analytical standard
C8H6Cl2O3
D7299
2,4-Dichlorophenoxyacetic acid, plant cell culture tested, ≥98%, crystalline
C8H6Cl2O3
D70724
2,4-Dichlorophenoxyacetic acid, 97%
C8H6Cl2O3
442840
4,4′-Isopropylidenediphenol, analytical standard
C15H16O2
A8380
5α-Androstan-17β-ol-3-one, ≥97.5%
C19H30O2
45726
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
C2H4O2
W200603
Acetic acid, natural, ≥99.5%, FG
C2H4O2
W200611
Acetic acid, ≥99.5%, FCC, FG
C2H4O2
ARK2183
Acetic acid, glacial, PharmaGrade, USP, JP, Ph Eur, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production.
C2H4O2
606766
Acetic acid-12C2, 99.9 atom % 12C
12C2H4O2
49085
Atrazine, analytical standard
C8H14ClN5
48187
Atrazine solution, certified reference material, 1000 μg/mL in methyl tert-butyl ether
C8H14ClN5
239658
Bisphenol A, ≥99%
C15H16O2
133027
Bisphenol A, 97%
C15H16O2
426008
Carbofuran, 98%
C12H15NO3
PS754
Carbofuran, analytical standard
C12H15NO3
542776
Ethyl (2R)-(+)-2-[4-(6-chlorobenzoxazol-2-yloxy)phenoxy]propionate, 95%
C18H16ClNO5
A9967
Glacial acetic acid, meets USP testing specifications
C2H4O2
108154
Hexyl acetate, 99%
C8H16O2
W256501
Hexyl acetate, ≥98%, FCC, FG
C8H16O2
W256528
Hexyl acetate, natural, ≥98%, FCC, FG
C8H16O2
746452
Magnesium sulfate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
MgSO4
793612
Magnesium sulfate, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥97%
MgSO4
M2643
Magnesium sulfate, BioReagent, suitable for cell culture, suitable for insect cell culture
MgSO4
203726
Magnesium sulfate, ≥99.99% trace metals basis
MgSO4
M3409
Magnesium sulfate solution, for molecular biology, 1.00 M±0.04 M
MgSO4
83266
Magnesium sulfate solution, BioUltra, for molecular biology
MgSO4