EMAIL THIS PAGE TO A FRIEND
Talanta
Thermally-initiated free radical polymerization for reproducible production of stable linear polyacrylamide coated capillaries, and their application to proteomic analysis using capillary zone electrophoresis-mass spectrometry.
PMID 26695337