EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Activation of AMP-activated protein kinase decreases receptor activator of NF-κB ligand expression and increases sclerostin expression by inhibiting the mevalonate pathway in osteocytic MLO-Y4 cells.
PMID 26713363
Abstract
AMP-activated protein kinase (AMPK) plays important roles in bone metabolism; however, little is known about its role in osteocytes. This study investigated the effects of AMPK activation on the expression of receptor activator of NF-κB ligand (RANKL) and sclerostin in osteocytes. Real-time PCR showed that AMPK activation by 5-aminoimidazole-4-carboxamide ribonucleotide (AICAR) significantly decreased the expression of Rankl in a dose- and time-dependent manner and significantly increased the expression of Sost, the gene encoding sclerostin, in osteocytic MLO-Y4 cells. Western blotting confirmed that AICAR decreased RANKL protein levels and increased sclerostin levels. In addition, suppression of AMPKα1 by siRNA significantly increased the expression of Rankl on 4 days after the transfection of siRNA, while Sost expression was not changed. Simvastatin, an inhibitor of HMG-CoA reductase, significantly decreased Rankl expression and increased Sost expression in MLO-Y4 cells. Supplementation with mevalonate or geranylgeranyl pyrophosphate, which are downstream metabolites of HMG-CoA reductase, significantly reversed the effects of AICAR. These findings indicated that AMPK regulated RANKL and sclerostin expression through the mevalonate pathway in osteocytes.