EMAIL THIS PAGE TO A FRIEND
Lymphology
A NOVEL FLT4 GENE MUTATION AND MR LYMPHANGIOGRAPHY IN A CHINESE FAMILY WITH MILROY DISEASE.
PMID 26714373
Abstract
Milroy disease is a congenital onset lymphedema linked to FLT4 gene mutations in the tyrosine kinase domain. So far, a total of 59 different FLT4 variants have been identified. Here, we report a novel FLT4 gene mutation in a Chinese family with Milroy disease and present their clinical symptoms and MR lymphangiographic findings.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
CKOZFN1023 CompoZr® Knockout ZFN Kit, human FLT4 (NM_002020)