EMAIL THIS PAGE TO A FRIEND
Oncotarget
EGFRvIII/integrin β3 interaction in hypoxic and vitronectinenriching microenvironment promote GBM progression and metastasis.
PMID 26717039
Abstract
Glioblastoma (GBM) is one of the most lethal brain tumors with a short survival time. EGFR amplification and mutation is the most significant genetic signature in GBM. About half of the GBMs with EGFR amplification express a constitutively autophosphorylated variant of EGFR, known as EGFRvIII. Our in vitro data demonstrated further enhanced EGFRvIII activity and tumor cell invasion in the tumor microenvironment of hypoxia plus extracellular matrix (ECM) vitronectin, in which EGFRvIII and integrin β3 tended to form complexes. The treatment with ITGB3 siRNA or the integrin antagonist cilengetide preferentially interrupted the EGFRvIII/integrin β3 complex, effectively reduced tumor cell invasion and activation of downstream signaling effectors. Cilengitide is recently failed in Phase III CENTRIC trial in unselected patients with GBM. However, we found that cilengitide demonstrated efficacious tumor regression via inhibition of tumor growth and angiogenesis in EGFRvIII orthotopic xenografts. Bioinformatics analysis emphasized key roles of integrin β3, hypoxia and vitronectin and their strong correlations with EGFRvIII expression in malignant glioma patient samples in vivo. In conclusion, we demonstrate that EGFRvIII/integrin β3 complexes promote GBM progression and metastasis in the environment of hypoxia and vitronectin-enrichment, and cilengitide may serve as a promising therapeutics for EGFRvIII-positive GBMs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EMU071351 MISSION® esiRNA, esiRNA targeting mouse Src (esiRNA1)