EMAIL THIS PAGE TO A FRIEND
Oncology letters
Low-dose interleukin-8 induces the adhesion, migration and invasion of the gastric cancer SGC-7901 cell line.
PMID 26722255
Abstract
Interleukin-8 (IL-8), an important inflammatory cytokine, is strongly associated with gastric cancer development and metastasis. High-dose (>1 ng/ml) IL-8 has been revealed to promote the adhesion, migration and invasion of human gastric cancer SGC-7901 cells in a dose-dependent manner. However, the IL-8 level produced by gastric cells is marginal, at even less than 1 ng/ml. It is unclear whether low-dose IL-8 also induces these capacities. In the present study, the effect of low-dose IL-8 on the adhesion, migration and invasion of the SGC-7901 cell line and the underlying molecular mechanism with regard to cluster of differentiation 44 (CD44) were investigated. The SGC-7901 cells were exposed to various concentrations of IL-8 (0, 0.2, 0.5, 0.8 and 1 ng/ml)