EMAIL THIS PAGE TO A FRIEND
Indian journal of pharmacology
Metabolic effects as a cause of myotoxic effects of fluoroquinolones.
PMID 26729952
Abstract
To investigate if fluoroquinolones (FQs) influence skeletal muscle metabolism of healthy and malignant hyperthermia susceptible (MHS) pigs. After approval from of the Animal Care Committee, 10 MHS pigs, and 6 MHS pigs were anesthetized with hemodynamic and systemic metabolic monitoring. Microdialysis catheters were placed intramuscularly. After equilibration, levofloxacin and ciprofloxacin were injected as a rapid bolus and continuous infusions. Lactate was measured in the dialysate and statistically analyzed was done (Wilcoxon-test; U-test; P < 0.05). There were no differences in age, weight, and baseline lactate levels between the groups. Both applications of levofloxacin- and ciprofloxacin-induced an increase of local lactate levels in healthy and MHS pigs. No difference between the two groups was observed. FQs influence skeletal muscle metabolism. Myotoxic effects of FQs can, therefore, be explained by an influence on the cellular energy balance.