EMAIL THIS PAGE TO A FRIEND
Beilstein Journal of Nanotechnology
Fabrication of hybrid graphene oxide/polyelectrolyte capsules by means of layer-by-layer assembly on erythrocyte cell templates.
PMID 26734521
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
295132
Chlorine, ≥99.5%
Cl2