EMAIL THIS PAGE TO A FRIEND
Clinical chemistry
Automated assay specific for cyclosporine in whole blood, with use of a chromium dioxide particle column.
PMID 2673583
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
480037
Magtrieve
CrO2