EMAIL THIS PAGE TO A FRIEND
Cellular and molecular gastroenterology and hepatology
Spontaneous Pancreatitis Caused by Tissue-Specific Gene Ablation of
PMID 26740970
Abstract
Perturbations in pancreatic ductal bicarbonate secretion cause chronic pancreatitis. The physiologic mechanism of ductal secretion is known, but its transcriptional control is not. We determine the role of the transcription factor hematopoietically expressed homeobox protein ( We derived mice with pancreas-specific, Cremediated Although