EMAIL THIS PAGE TO A FRIEND
Molecular neurobiology
Inhibitory Effect of Endomorphin-2 Binding to the μ-Opioid Receptor in the Rat Pre-Bötzinger Complex on the Breathing Activity.
PMID 26742523
Abstract
Opiates are commonly used analgesics that often cause clinical respiratory depression. However, their underlying mechanisms remain unclear. Endomorphin-2 (EM2) is a novel, endogenous tetrapeptide opioid with very high affinity and selectivity for the μ-opioid receptor (MOR). The pre-Bötzinger complex (pre-BötC) is considered the center of respiratory rhythm generation, and the synaptic connections in this region are essential for respiratory rhythm. The present study identified EM2-like immunoreactive (LI) axonal terminals in the pre-BötC of adult rats. Some EM2-LI axonal terminals made principally symmetric synapses with neurokinin 1 receptor (NK1R)-LI or MOR-LI neuronal dendritic processes in the pre-BötC. Unilateral microinjection of EM2 into the pre-BötC decreased breathing frequency and amplitude. A prior microinjection of the selective MOR antagonist β-funaltrexamine (β-FNA) into the pre-BötC prevented the effects of EM2. The present results suggest that EM2-LI axonal terminals modulate NK1R-expressing neurons in the pre-BötC and that EM2 plays a role in respiratory depression through MORs in the pre-BötC.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E7384
[D-Ala2, N-Me-Phe4, Gly5-ol]-Enkephalin acetate salt, ≥97% (HPLC)
C26H35N5O6