EMAIL THIS PAGE TO A FRIEND
Natural Product Communications
Antioxidant and Anti-inflammatory Activities of Essential Oil and Extracts of Piper miniatum.
PMID 26749849
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
127914
1-Phenyl-3-pyrazolidinone, 97%
C9H10N2O