EMAIL THIS PAGE TO A FRIEND
Quarterly reviews of biophysics
Antitumour drug-DNA interactions: NMR studies of echinomycin and chromomycin complexes.
PMID 2675172
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0477 Echinomycin, ≥98% (HPLC)
C51H64N12O12S2