EMAIL THIS PAGE TO A FRIEND
Nature communications
Enhanced electronic properties in mesoporous TiO2 via lithium doping for high-efficiency perovskite solar cells.
PMID 26758549
Abstract
Perovskite solar cells are one of the most promising photovoltaic technologies with their extraordinary progress in efficiency and the simple processes required to produce them. However, the frequent presence of a pronounced hysteresis in the current voltage characteristic of these devices arises concerns on the intrinsic stability of organo-metal halides, challenging the reliability of technology itself. Here, we show that n-doping of mesoporous TiO2 is accomplished by facile post treatment of the films with lithium salts. We demonstrate that the Li-doped TiO2 electrodes exhibit superior electronic properties, by reducing electronic trap states enabling faster electron transport. Perovskite solar cells prepared using the Li-doped films as scaffold to host the CH3NH3PbI3 light harvester produce substantially higher performances compared with undoped electrodes, improving the power conversion efficiency from 17 to over 19% with negligible hysteretic behaviour (lower than 0.3%).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
142379
4-tert-Butylpyridine, 98%
C9H13N