Turkish journal of medical sciences
Decreasing the need for mechanical ventilation after surgery for retinopathy of prematurity: sedoanalgesia vs. general anesthesia.
PMID 26775385
Premature infants experience more respiratory problems after surgical procedures. We aimed to compare general anesthesia with sedation regarding the need for postoperative mechanical ventilation in infants undergoing retinopathy of prematurity (ROP) surgery. Sixty patients who underwent laser surgery for ROP were included in this study. This study was performed between October 2010 and December 2012. The sedation group (Group S, n = 30) received 1 mg/kg ketamine and 1 mg/kg propofol as a bolus for induction. The patients then received an infusion of 100-150 μg kg(-1) min(-1) propofol and 0.25 mg kg(-1) h(-1) ketamine for maintenance. In the general anesthesia group (Group G, n = 30), anesthesia was induced using 8% sevoflurane by inhalation with 50% nitrous oxide in oxygen. Anesthesia was maintained with sevoflurane (2%) and 50% nitrous oxide in oxygen. There was no difference in gestational age, birth weight, current age, or current body weight between the two groups. Preoperative medical histories of the groups were similar. Two patients in Group S and 11 patients in Group G required postoperative mechanical ventilation (P = 0.010). Blood pressures and heart rates were similar. In premature infants, sedoanalgesia administration reduced the need for postoperative mechanical ventilation after surgery for ROP.
Related Materials
Product #
Molecular Formula
Add to Cart
Aminophylline, ≥98%, powder
C7H8N4O2 · 0.5C2H8N2