EMAIL THIS PAGE TO A FRIEND
Food chemistry
Localized lipid autoxidation initiated by two-photon irradiation within single oil droplets in oil-in-water emulsions.
PMID 26776033
Abstract
The initiation of lipid autoxidation within single oil droplets in Tween-20-stabilized oil-in-water emulsion was achieved by highly focused two-photon (2P) irradiation at excitation wavelength (λex) 700 nm. The radical formation was enhanced by inclusion of the photo-cleavable radical initiator di-tert-butyl peroxide (DTBP) into the droplets, and demonstrated with confocal microscopy using radical-sensitive probe BODIPY(665/676). The radical chain reactions progressed up to 60 μm; however, there were no indications of oxidation in neighboring droplets demonstrating that radicals and oxidized probe molecules were not able to migrate between oil droplets. In addition, the spatial propagation of lipid autoxidation increased with the degree of oil unsaturation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
168521
Luperox® DI, tert-Butyl peroxide, 98%
C8H18O2