EMAIL THIS PAGE TO A FRIEND
Food chemistry
Selective solid-phase extraction using molecularly imprinted polymer as a sorbent for the analysis of fenarimol in food samples.
PMID 26776046
Abstract
In the present communication, a non-covalent fenarimol-imprinted polymer was synthesized by precipitation polymerization technique using methacrylic acid (MAA) as a functional monomer, ethylene glycol dimethacrylate (EGDMA) as a cross-linker, and azobisisobutyronitrile (AIBN) as an initiator in different porogenic solvent. Binding study of molecularly imprinted and non-imprinted polymer (MIP and NIP) showed that MIP possesses a higher affinity towards this analyte compared to NIP. The binding affinity of MIP was calculated by static and kinetic adsorption study. Further, a MIP based cartridge was designed to use in extraction process, necessary for specific determination and quantification of the fungicide in food matrices. Under the optimum conditions, developed method was found to be linear (R(2)=0.9999-0.9994). Limit of detection (LOD) and limit of quantitation (LOQ) in samples were 0.03-0.06 and 0.12-0.21 μg mL(-1), respectively. The rate of recovery of fenarimol was 91.16-99.52% on MIPs. The validated method of molecularly imprinted solid-phase extraction (MISPE) cartridge was successfully applied to the food matrices and compared with commercial sorbent (RP18 and Oasis HLB). However we feel, this method has promising applications in the routine analysis of food samples in industry.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45478
Ethirimol, PESTANAL®, analytical standard
C11H19N3O
45484
Fenarimol, PESTANAL®, analytical standard
C17H12Cl2N2O