EMAIL THIS PAGE TO A FRIEND
Oncology reports
Activation of NLRP3 inflammasome enhances the proliferation and migration of A549 lung cancer cells.
PMID 26782741
Abstract
Lung cancer is the leading cause of cancer death, and it is widely accepted that chronic inflammation is an important risk for the development of lung cancer. Now, it is recognized that the nucleotide-binding and oligomerization domain (NOD) like receptors (NLRs)-containing inflammasomes are involved in cancer-related inflammation. This study was designed to investigate the effects of NLR family pyrin domain containing protein 3 (NLRP3) inflammasome on the proliferation and migration of lung adenocarcinoma cell line A549. Using 5-ethynyl-2'-deoxyuridine (EdU) incorporation assay, scratch assay, and Transwell migration assay, we showed that activation of the NLRP3 inflammasome by LPS+ATP enhanced the proliferation and migration of A549 cells. Western blot analysis showed that activation of phosphorylation of Akt, ERK1/2, CREB and the expression of Snail increased, while the expression of E-cadherin decreased after the activation of NLRP3 inflammasome. Moreover, these effects were inhibited by the following treatments: i) downregulating the expression of NLRP3 by short hairpin RNA (shRNA) interference, ii) inhibiting the activation of NLRP3 inflammasome with a caspase-1 inhibitor, iii) blocking the interleukin-1β (IL-1β) and IL-18 signal transduction with IL-1 receptor antagonist (IL-1Ra) and IL-18 binding protein (IL-18BP). Collectively, these results indicate that NLRP3 inflammasome plays a vital role in regulating the proliferation and migration of A549 cells and it might be a potential target for the treatment of lung cancer.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EHU071121 MISSION® esiRNA, esiRNA human NLRP3 (esiRNA1)