EMAIL THIS PAGE TO A FRIEND
Journal of food science and technology
Vegetable product containing caseinomacropeptide and germinated seed and sprouts.
PMID 26788011
Abstract
In this study vegetable product containing germinated seed and sprouts of lentils and cowpeas, and caseinomacropeptide isolated from whey is produced. Three different forms of vegetable product namely puree (VP), freeze-dried (FD) and drum-dried (DD) are produced. Freeze-dried and DD forms are produced to diversify forms of utilization and to improve functionality such as increased shelf life and decreased storage space. Their beneficial effects on health are determined using in vitro methods. All forms displayed antioxidant activities against DPPH radical and oxygen radical, α-amylase inhibitory activities, bile acid binding capacities, and angiotension converting enzyme (ACE) inhibitory activities. Freeze-dried product exhibited the strongest inhibition on α-amylase and ACE with the IC50 value 0.09 μM total phenolic and 0.82 mg protein/g sample, respectively when evaluated on the basis of serving size.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G2878
Glycocholic acid hydrate, synthetic, ≥97% (HPLC)
C26H43NO6 · xH2O