EMAIL THIS PAGE TO A FRIEND
Water research
Low pressure UV/H2O2 treatment for the degradation of the pesticides metaldehyde, clopyralid and mecoprop - Kinetics and reaction product formation.
PMID 26803264
Abstract
The degradation kinetics of three pesticides - metaldehyde, clopyralid and mecoprop - by ultraviolet photolysis and hydroxyl radical oxidation by low pressure ultraviolet hydrogen peroxide (LP-UV/H2O2) advanced oxidation was determined. Mecoprop was susceptible to both LP-UV photolysis and hydroxyl radical oxidation, and exhibited the fastest degradation kinetics, achieving 99.6% (2.4-log) degradation with a UV fluence of 800xa0mJ/cm(2) and 5xa0mg/L hydrogen peroxide. Metaldehyde was poorly degraded by LP-UV photolysis while 97.7% (1.6-log) degradation was achieved with LP-UV/H2O2 treatment at the maximum tested UV fluence of 1000xa0mJ/cm(2) and 15xa0mg/L hydrogen peroxide. Clopyralid was hardly susceptible to LP-UV photolysis and exhibited the lowest degradation by LP-UV/H2O2 among the three pesticides. The second-order reaction rate constants for the reactions between the pesticides and OH-radicals were calculated applying a kinetic model for LP-UV/H2O2 treatment to be 3.6xa0×xa010(8), 2.0xa0×xa010(8) and 1.1xa0×xa010(9)xa0M(-1)xa0s(-1) for metaldehyde, clopyralid and mecoprop, respectively. The main LP-UV photolysis reaction product from mecoprop was 2-(4-hydroxy-2-methylphenoxy) propanoic acid, while photo-oxidation by LP-UV/H2O2 treatment formed several oxidation products. The photo-oxidation of clopyralid involved either hydroxylation or dechlorination of the ring, while metaldehyde underwent hydroxylation and produced acetic acid as a major end product. Based on the findings, degradation pathways for the three pesticides by LP-UV/H2O2 treatment were proposed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
36758
Clopyralid, PESTANAL®, analytical standard
C6H3Cl2NO2