EMAIL THIS PAGE TO A FRIEND
Enfermedades infecciosas y microbiologia clinica
Galactomannan enzyme immunoassay and quantitative Real Time PCR as tools to evaluate the exposure and response in a rat model of aspergillosis after posaconazole prophylaxis.
PMID 26804792
Abstract
A steroid-immunosuppressed rat model of invasive pulmonary aspergillosis was use to examine the usefulness of galactomannan enzyme immunoassay (GM) and quantitative real time PCR (RT-PCR) in evaluating the association between response and exposure after a high dose of prophylactic posaconazole. Two different strains of Aspergillus fumigatus with different in vitro posaconazole susceptibility were used. Serum concentrations demonstrated similar posaconazole exposure for all treated animals. However, response to posaconazole relied on the in vitro susceptibility of the infecting strain. After prophylaxis, galactomannan index and fungal burden only decreased in those animals infected with the most susceptible strain. This study demonstrated that both biomarkers may be useful tools for predicting efficacy of antifungal compounds in prophylaxis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
32103
Posaconazole, VETRANAL, analytical standard
C37H42F2N8O4