EMAIL THIS PAGE TO A FRIEND
AIDS (London, England)
Mitochondrial injury and cognitive function in HIV infection and methamphetamine use.
PMID 26807965
Abstract
In this work, we evaluated the association of human immunodeficiency virus (HIV) infection and methamphetamine (METH) use with mitochondrial injury in the brain and its implication on neurocognitive impairment. Mitochondria carry their genome (mtDNA) and play a critical role in cellular processes in the central nervous system. METH is commonly used in HIV-infected populations. HIV infection and METH use can cause damage to mtDNA and lead to neurocognitive morbidity. We evaluated HIV infection and METH use with mitochondrial injury in the brain. We obtained white and gray matter from Brodmann areas 7, 8, 9, 46 of the following: HIV-infected individuals with history of past METH use (HIV+METH+, n = 16), HIV-infected individuals with no history of past METH use (HIV+METH-, n = 11), and HIV-negative controls (HIV-METH-, n = 30). We used the 'common deletion', a 4977 bp mutation, as a measurement of mitochondrial injury, and quantified levels of mtDNA and 'common deletion' by droplet digital PCR, and evaluated in relation to neurocognitive functioning [Global Deficit Score (GDS)]. Levels of mtDNA and mitochondrial injury were highest in white matter of Brodmann area 46. A higher relative proportion of mtDNA carrying the 'common deletion' was associated with lower GDS (P < 0.01) in HIV+METH+ but higher GDS (P < 0.01) in HIV+METH-. Increased mitochondrial injury was associated with worse neurocognitive function in HIV+METH- individuals. Among HIV+METH+ individuals, an opposite effect was seen.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
80666
Hexarelin, ≥90% (HPLC)
C47H58N12O6