EMAIL THIS PAGE TO A FRIEND
Pesticide biochemistry and physiology
The subunit gene Ldα1 of nicotinic acetylcholine receptors plays important roles in the toxicity of imidacloprid and thiamethoxam against Leptinotarsa decemlineata.
PMID 26821658
Abstract
Nicotinic acetylcholine receptors (nAChRs) are pentameric ACh-gated ion channels. It is believed that nAChRs composed of different subunits may vary in their function and toxicological characteristics. Neonicotinoids are activators of nAChRs and important insecticides that are extensively used for crop protection and resistance has been developed by some pests. They are also major insecticides for the control of Leptinotarsa decemlineata, which is a destructive defoliator pest that invaded the Xinjiang region of China in the 1990s. However, little is known about the constitution or subunits of the target in this pest. In this study, the full-length cDNAs encoding four new nAChR subunits (named Ldα3, Ldα6, Ldα10, and Ldβ1) were cloned from L. decemlineata. These genes encode 822-, 753-, 672-, and 759-amino acid proteins, respectively, which share typical features of insect nAChRs subunits and closely resemble the corresponding subunits of the nAChRs from Tribolium castaneum. Temporal and spatial expression analyses showed that these genes, as well as the previously identified Ldα1, Ldα2, and Ldα8 genes, are widely expressed in all developmental stages, including eggs, larvae of various instars, pupae, and adults. All genes monitored were expressed at higher levels in the head than in the thorax and abdomen, except for Ldα10. Dietary ingestion of double-stranded RNA bacterially expressed for Ldα1 (dsLdα1) significantly reduced the mRNA level of Ldα1 in treated larvae and adults by 48.0% and 78.6%, respectively. Among the non-target genes, Ldα3, Ldα9, and Ldβ1 were significantly up-regulated in larvae. A toxicity bioassay showed that dsLdα1 treatment greatly decreased the sensitivity to imidacloprid and thiamethoxam in adults. The larval susceptibility to thiamethoxam but not to imidacloprid was also reduced because of the lower down-regulation of Ldα1. Thus, our results suggest that Ldα1 encodes a subunit of a functional nAChR that mediates the toxicity of imidacloprid and thiamethoxam against L. decemlineata and that the down-regulation of Ldα1 might be an important mechanism for resistance and/or tolerance of L. decemlineata to neonicotinoids.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
37894
Imidacloprid, PESTANAL®, analytical standard
C9H10ClN5O2
37924
Thiamethoxam, PESTANAL®, analytical standard
C8H10ClN5O3S