EMAIL THIS PAGE TO A FRIEND
Revista medico-chirurgicala a Societatii de Medici si Naturalisti din Iasi
[Pyridazinilides (XIV). The action of p-phenyl-phenacyl bromide on 3-phenyl-pyridazine].
PMID 2682894
Abstract
This time the synthesis of some new derivatives obtained by the quaternary substitution of one of the heterocyclic nitrogen atoms and by reactions of dipolar cycloaddition is presented. The results of the tests on their antimicrobial and antiinflammatory actions are discussed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
101087
2-Bromo-4′-phenylacetophenone, technical grade
C14H11BrO