EMAIL THIS PAGE TO A FRIEND
Journal of biomaterials science. Polymer edition
Mechanical and biological properties of photocurable oligolactide-HA composites investigated under accelerated degradation.
PMID 26838814
Abstract
The major concern related to biodegradable bone substitute materials is the loss of mechanical strength which can be undesirable when occurring too quickly before new bone formation. In this study, the multifunctional lactide oligomers having 2, 3, and 4 arms end capped with methacrylate groups were synthesized with the aim of improving the degradation properties. Their composites with hydroxyapatite (HA) were photopolymerized and subjected to accelerated degradation at 60 °C. The results showed that increasing number of arms significantly improved thermal and mechanical properties as well as biocompatibility of the composites. All composites although varying in number of arms had similar levels of bone-specific gene expression and calcification indicating their equal bioactivity in supporting bone formation. The high HA content in the composites was proposed to be responsible for enhanced osteoblast response, and this tended to suppress the effects of polymeric structure.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
276685
Methacrylic anhydride, contains 2,000 ppm topanol A as inhibitor, 94%
C8H10O3