EMAIL THIS PAGE TO A FRIEND
Materials science & engineering. C, Materials for biological applications
Characterization and application of roxithromycin loaded cyclodextrin based nanoparticles for treatment of multidrug resistant bacteria.
PMID 26838816
Abstract
An outbreak of infections with a high mortality rate caused by multidrug resistant (MDR) bacteria is one of the biggest health challenges globally. A class IV drug, roxithromycin (ROX), has poor solubility. In this study, ROX was first encapsulated in the cavity of each of the β-cyclodextrin (βCD) and hydroxypropyl-β-cyclodextrin (HPβCD). Then, each of the resulting βCD-ROX inclusion complex and HPβCD-ROX inclusion complex were separately loaded into poly-(lactic-co-glycolic acid) (PLGA) to synthesize βCD-ROX/PLGA and HPβCD-ROX/PLGA nanoparticles (NPs). Blank and ROX loaded PLGA (ROX-PLGA) NPs were also prepared. The loading efficiency of ROX is comparatively high for HPβCD-ROX/PLGA NPs in comparison to the βCD-ROX/PLGA NPs and ROX-PLGA NPs. All designed formulations showed significant (P<0.0001) antibacterial activity against the selected MDR bacterial strains. In a nutshell, this study demonstrated a great therapeutic potential of the above-mentioned delivery systems for treatment of MDR bacteria.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
303143
3,6-Dimethyl-1,4-dioxane-2,5-dione, 99%
C6H8O4
G1796
Glycolide, ≥99%
C4H4O4