EMAIL THIS PAGE TO A FRIEND
Anticancer research
Synergisitic and Antagonistic AML Cell Type-specific Responses to 5-Aza-2-deoxycitidine and 1-h-D-Arabinofuranoside.
PMID 26851026
Abstract
The search for synergistic drug combinations is critical to the treatment of drug-resistant cancer, such as acute myeloid leukemia (AML). Characterizing RNA expression associated with 5-aza-2'-deoxycytidine (DAC) and 1-h-D-arabinofuranosylcytosine (Ara-C) is a critical step to increase the efficacy of their combinatorial therapies. After 72 h of single-dose treatments of AML cells with DAC or Ara-C, the half-maximal effective concentration of DAC and Ara-C and the drug combination index were assessed. Pre-treatment with DAC restores cellular sensitivity in Ara-C-resistant AML cells. In contrast, DAC/Ara-C combinations are antagonistic in other Ara-C-sensitive AML cells. Our results provide an alternative approach for predicting what combinations, dosing and scheduling of drug delivery should be used to better individualize therapy of AML.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
360953
α,α-Dicyanoethyl acetate, 98%
C6H6N2O2